Yamato 241084 Solid Basket for Vertical Loading Sterilizers f/10771-series
Representative image only.
Yamato 241084 Solid Basket for Vertical Loading Sterilizers f/10771-series
Yamato | Mfr: 241084| Item: 1077151
Use these accessories for easier load organization
₹77,100.00INR / Each
₹77,100.00INR / Each
Product Description
Key Features
  • Use with Yamato Vertical Loading Sterilizers 10773-02, 10773-03, 10773-04, and 10773-05
More About this Item
Stainless steel solid basket is used for liquid-filled media and petri dishes, as well as to contain potential spillage from waste bags. It has a hinged lid with a handle. the side and bottom of the solid basket are perforated for better steam penetration.
Specifications
  • DescriptionSolid Basket for Vertical Loading Sterilizers f/10771-series