WTW 301972 TetraCon 4-electrode conductivity probe, 10-m cable, waterproof DIN connector
Representative image only.
WTW 301972 TetraCon 4-electrode conductivity probe, 10-m cable, waterproof DIN connector
WTW | Mfr: 301972| Item: 1970782
Four-electrode conductivity cells offer a high degree of accuracy over wide measuring range
₹256,000.00INR / Each
₹256,000.00INR / Each
Buy It Together
Current Item
WTW 301972 TetraCon 4-electrode conductivity probe, 10-m cable, waterproof DIN connector
WTW 301972 TetraCon 4-electrode conductivity probe, 10-m cable, waterproof DIN connector
1970782
₹256,000.00INR / Each
+ =
Total Price 522000.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Do not need platinization
  • 3 year warranty
More About this Item
WTW conductivity probes to suit your conductivity measurement needs.
3 Year Warranty
Specifications
  • Accessory TypeK=0.475
  • Length (in)393
  • Description4-electrode conductivity probe, 10-m cable, waterproof DIN connector