WTW 301961 LR 325/01 conductivity probe (Ultra-pure water)
Representative image only.
WTW 301961 LR 325/01 conductivity probe (Ultra-pure water)
WTW | Mfr: 301961| Item: 1970662
Four-electrode conductivity cells offer a high degree of accuracy over wide measuring range
₹282,000.00INR / Each
₹282,000.00INR / Each
Buy It Together
Current Item
WTW 301961 LR 325/01 conductivity probe (Ultra-pure water)
WTW 301961 LR 325/01 conductivity probe (Ultra-pure water)
1970662
₹282,000.00INR / Each
+ =
Total Price 548000.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Do not need platinization
  • 3 year warranty
More About this Item
WTW conductivity probes to suit your conductivity measurement needs.
3 Year Warranty
Specifications
  • Accessory TypeK=0.1
  • Length (in)39
  • DescriptionTetraCon 4-electrode conductivity probe, 3-m cable, waterproof DIN connector