WTW 285126293 Conductivity Standard, 1.41/12.9/112 mS; 3 x 6 Ampoules Ea

Representative image only.

WTW 285126293 Conductivity Standard, 1.41/12.9/112 mS; 3 x 6 Ampoules Ea

WTW | Mfr: 285126293| Item: 5935146

Offering up to 5 years shelf life

₹34,700.00INR / Pkg of 18

₹34,700.00INR / Pkg of 18
Product Description

Key Features

  • Highly precise standards with ±0.25% accuracy
  • Certified accuracy traceable to NIST/DIN standards
  • Single-use buffers with 25-mL of solution
  • 2 year warranty

More About this Item

Presterilized and free of preservatives, these ready-to-use, sealed glass ampoules of conductivity standards offer a 5-year shelf life. 25 mL solution per ampoule.

Six ampoules of each of the 3 standard values.

Two year warrantyConformite Europeenne
Specifications
  • Volume (mL)25
  • Volume (oz)0.9
  • Volume3x 6 Ampoules each
  • DescriptionConductivity Standard, 1.41/12.9/112 mS; 3 x 6 Ampoules Ea
  • Warranty2 year
Reviews
Q&A