Worm Feed Auger for Wet or Oily Grinding
Representative image only.
Worm Feed Auger for Wet or Oily Grinding
| Mfr: S-E02-1/2W| Item: 0418656
Mills feeders for dry and wet grinding
₹26,600.00INR / Each
₹26,600.00INR / Each
Product Description
Key Features
  • Use with wet or oily materials
More About this Item
Select either tooth feed auger for dry grinding or worm feed auger for wet or oily grinding.
Specifications
  • Compatible WithPlate mills 04186-00 and -05
  • ApplicationWet or oily grinding
  • DescriptionWorm Feed Auger for Wet or Oily Grinding