Wiretrol II Capillary Micropipettes, Glass, 1-5 µL; 250/Pk
Representative image only.
Wiretrol II Capillary Micropipettes, Glass, 1-5 µL; 250/Pk
Wiretrol | Mfr: 5-000-2005| Item: 0795100
Micropipettes provide accuracy and convenience with dual graduations
₹13,400.00INR / Pkg of 250
₹13,400.00INR / Pkg of 250
Product Description
Includes
short plunger.
Key Features
  • Dual graduations for noncontact delivery
  • ±1% accuracy
  • 3 month warranty
More About this Item
Draw fluid into the calibrated glass barrel with fingertip plunger. Accuracy is ±1% of contained volume. Each pack contains 250 disposable glass barrels and two plungers: use long plunger to clear barrel; use short plunger to maintain air cushion between plunger and sample.
3 Month Warranty
Specifications
  • SterileNo
  • Max Volume (µL)5
  • Min Volume (µL)1
  • Accuracy (%)±1.00%
  • Overall Length (InchShort)3.5
  • Overall Length (mm)90
  • Wetted MatterialGlass
  • Color CodeWhite
  • DescriptionPositive Displacement Microdispensers, 1-5 uL; 250/pk