Winland Electronics EAPRO-WTS EnviroAlert Professional® Wireless Temperature Sensor, 32° to 122° F (0° to 50° C)
Representative image only.
Winland Electronics EAPRO-WTS EnviroAlert Professional® Wireless Temperature Sensor, 32° to 122° F (0° to 50° C)
Winland Electronics | Mfr: M-001-0153| Item: 1045007
Monitoring made easy with wireless sensors
₹74,800.00INR / Each
₹74,800.00INR / Each
Product Description
Key Features
  • Measure ambient room conditions
  • Indoor transmission distance of 500’ (152.4 m) - (results may vary)
  • Set temperature high/low set points on EAPro-Gateway device
  • Update remotely with INSIGHT (INSIGHT sold separately)
More About this Item
For use with Winland Electronics EnviroAlert Professional® Gateway.
Do NOT put EAPro-GTWY or EAPro-WTS devices into cooler/freezer environment. Use a hard-wired sensor with EAProGTWY or EAPro-WMFT. The EAPro-WTS is NOT a standalone device.
1 Year WarrantyConformite Europeenne
Specifications
  • Compatible WithWinland Electronics EAPRO-GTWY EnviroAlert Professional® Gateway
  • DescriptionEnviroAlert Professional® Wireless Temperature Sensor, 32° to 122° F (0° to 50° C)