Winland Electronics EAPRO-WMFT EnviroAlert Professional® Wireless Multi-Function Transmitter
Representative image only.
Winland Electronics EAPRO-WMFT EnviroAlert Professional® Wireless Multi-Function Transmitter
Winland Electronics | Mfr: M-001-0155| Item: 1045009
Monitoring made easy with wireless sensors
₹73,700.00INR / Each
₹73,700.00INR / Each
Product Description
Key Features
  • Measure ambient room conditions
  • The EAPro-WMFT requires an accessory sensor for operation
  • Monitoring for temperature, presence of water and more
  • Max sensor cable run of 1,000’ (304.8 m) [18-22 twisted pair]
  • Indoor transmission distance of 500’ (152.4 m) - (results may vary)
  • Set humidity high/low set points on EAPro-Gateway device
  • Update remotely with INSIGHT (INSIGHT sold separately)
More About this Item
For use with Winland Electronics EnviroAlert Professional® Gateway.
Do NOT put EAPro-GTWY or EAPro-WMFT devices into cooler/freezer environment. Use a hard-wired sensor with EAProGTWY or EAPro-WMFT. The EAPro-WMFT is NOT a standalone device.
1 Year WarrantyConformite Europeenne
Specifications
  • Compatible WithWinland Electronics EAPRO-GTWY EnviroAlert Professional® Gateway
  • DescriptionEnviroAlert Professional® Wireless Multi-Function Transmitter