Winland Electronics EAPRO-WHS EnviroAlert Professional® Wireless Humidity Sensor, 5 to 95% RH
Representative image only.
Winland Electronics EAPRO-WHS EnviroAlert Professional® Wireless Humidity Sensor, 5 to 95% RH
Winland Electronics | Mfr: M-001-0154| Item: 1045008
Monitoring made easy with wireless sensors
₹78,300.00INR / Each
₹78,300.00INR / Each
Product Description
Key Features
  • Measure ambient room conditions
  • Wireless monitoring of ambient room humidity
  • Indoor transmission distance of 500’ (152.4 m) - (results may vary)
  • Set humidity high/low set points on EAPro-Gateway device
  • Update remotely with INSIGHT (INSIGHT sold separately)
More About this Item
For use with Winland Electronics EnviroAlert Professional® Gateway.
Do NOT put EAPro-GTWY or EAPro-WHS devices into cooler/freezer environment. Use HA-4 inside cooler/freezer hard wired to an EAPro-GTWY. The EAPro-WHS is NOT a standalone device.
1 Year WarrantyConformite Europeenne
Specifications
  • Compatible WithWinland Electronics EAPRO-GTWY EnviroAlert Professional® Gateway
  • DescriptionEnviroAlert Professional® Wireless Humidity Sensor, 5 to 95% RH