Wildco 40-A50 Wisconsin Plankton net sampler with case
Representative image only.
Wildco 40-A50 Wisconsin Plankton net sampler with case
Wildco | Mfr: 40-A50| Item: 0549100
Two-hoop, 80-µm nylon net support system
₹129,000.00INR / Each
₹129,000.00INR / Each
Product Description
Key Features
  • Tapered SS bucket with 9-in2 filter area
  • 3 month warranty
More About this Item
Use for vertical sampling and for obtaining plankton from streams, rivers, and lakes. Bucket screws onto sampler and has a drain area with hose and hose clamp to simplify sample retrieval. Sampler measures 30"L x 7" dia; mouth diameter is 5". Includes case.
3 Month Warranty
Specifications
  • MaterialStainless Steel
  • DescriptionWisconsin Plankton net sampler with case