Welch 1420H-14 Foreline dry ice cold trap, 1/2" OD straight tube connection
Representative image only.
Welch 1420H-14 Foreline dry ice cold trap, 1/2" OD straight tube connection
Welch | Mfr: 1420H-14| Item: 7920930
₹400,000.00INR / Each
₹400,000.00INR / Each
Product Description
Key Features
  • Protect vacuum pump against damages from contaminants
  • 1 year warranty
More About this Item
These traps are easy to clean—remove from the vacuum pump and clean at the end of each day to prevent trapped oil vapors from damaging the pump. Dry ice traps have a removable 3-qt well for dry ice/alcohol slurry. Liquid nitrogen cold traps efficiently trap acidic vapors and lower the vacuum pump base pressure.
1 Year Warranty
Specifications
  • Length (InchShort)10
  • Height (in)8.25
  • Diameter (InchShort)10
  • DescriptionForeline dry ice cold trap, 1/2" OD straight tube connection