Watson-Marlow 520Ah Handswitch
Representative image only.
Watson-Marlow 520Ah Handswitch
Watson-Marlow | Mfr: 059.3022.000| Item: 5013507
Enhance your Watson-Marlow peristaltic pump operations with specialized control accessories, designed for precision and ease of use in demanding laboratory and industrial environments
₹33,400.00INR / Each
₹33,400.00INR / Each
Buy It Together
Current Item
Watson-Marlow 520Ah Handswitch
Watson-Marlow 520Ah Handswitch
5013507
₹33,400.00INR / Each
+ + + =
Total Price 3254400.00 ADD 4 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Equipped with a 25-pin D-connector, connect directly to models 323Du and 520Du for seamless integration
  • Use the footswitch for hands-free operation that allows for multitasking and maintains sterile conditions
  • Use the handswitch for precise manual control with a comfortable ergonomic design
More About this Item
Optimize your fluid handling processes with these essential accessories, ensuring enhanced control and efficiency in your applications. Both the hand and footswitch will operate the MemoDose function on Watson-Marlow pumps 323Du and 520Du.
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
Specifications
  • Compatible WithWatson-Marlow 323Du and 520Du pumps
  • DescriptionHandswitch