W.S. Tyler Sieve, Half Height, Brass, 200 mm OD, 0.600 mm Openings, No. 30 mesh
Representative image only.
W.S. Tyler Sieve, Half Height, Brass, 200 mm OD, 0.600 mm Openings, No. 30 mesh
W.S. Tyler | Mfr: 9626| Item: 5994316
Repeatable results and particle size distribution
₹18,300.00INR / Each
₹18,300.00INR / Each
Buy It Together
Current Item
W.S. Tyler Sieve, Half Height, Brass, 200 mm OD, 0.600 mm Openings, No. 30 mesh
W.S. Tyler Sieve, Half Height, Brass, 200 mm OD, 0.600 mm Openings, No. 30 mesh
5994316
₹18,300.00INR / Each
+ =
Total Price 36200.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Constructed with a brass frame and stainless steel wire
  • 1 year warranty
More About this Item
These half height brass sieves conform to current ISO 3310-1 and BS410 specifications.
1 Year Warranty
Specifications
  • MaterialBrass frame, stainless steel wire
  • U.S. Standard Screen Size (No.)30
  • Tyler Standard Screen Size (mesh)28
  • Sieve Opening (mm)0.595
  • Outside Diameter (in)8
  • Height (InchShort)1
  • DescriptionSieve, brass, 200 mm OD, 0.600 mm openings, half height, No. 30 mesh