Velp Test Tubes for Digestion System, 400 mL; 1/EA

Representative image only.

Velp Test Tubes for Digestion System, 400 mL; 1/EA

Velp | Mfr: A00000185| Item: 8658019

Expand the capabilities of your digestion system

₹28,500.00INR / Each

₹28,500.00INR / Each
Product Description

Key Features

  • Racks, caps, supports, stands, and more!
  • Compatible with Velp Models: DKL12 Test tube size: 400 mL
  • 2 year warranty

More About this Item

These accessories are compatible with Velp's DK and DKL Digestion Systerms.

Two year warranty
Specifications
  • Tube Size (mL)400
  • Materials of ConstructionGlass
  • DescriptionTest Tubes for Digestion System, 400 mL; 1/EA
Reviews
Q&A