Velp Test Tubes for Digestion System, 250 mL; 3/BX

Representative image only.

Velp Test Tubes for Digestion System, 250 mL; 3/BX

Velp | Mfr: A00000144| Item: 8658044

Expand the capabilities of your digestion system

₹35,000.00INR / Box of 3

₹35,000.00INR / Box of 3
Product Description

Key Features

  • Racks, caps, supports, stands, and more!
  • Compatible with Velp Models: DK6, DK8, DK20, DKL8, DKL12, and DKL20
  • 2 year warranty

More About this Item

These accessories are compatible with Velp's DK and DKL Digestion Systerms.

Two year warranty
Specifications
  • Tube Size (mL)250
  • Materials of ConstructionGlass
  • DescriptionTest Tubes for Digestion System, 250 mL; 3/BX
Reviews
Q&A