Velp Fp4-Portable FP4 Portable 4-position Flocculator 100-240V

Representative image only.

Velp Fp4-Portable FP4 Portable 4-position Flocculator 100-240V

Velp | Mfr: F105A0117| Item: 9956105

Save money and chemicals by testing on a small scale

₹237,210.00INR / Each

₹237,210.00INR / Each

Buy It Together

Current Item

Velp Fp4-Portable FP4 Portable 4-position Flocculator 100-240V
Velp Fp4-Portable FP4 Portable 4-position Flocculator 100-240V
9956105

₹237,210.00INR / Each
+ + =
Total Price 240610.00 ADD 3 ITEMS TO CART
Product Description

Includes

stainless steel stirring rods and universal AC adapter.

Key Features

  • ASTM D2035 compliant method
  • Single motor drives all positions at a consistent speed
  • Choose from portable or laboratory models, with four or six positions
  • 2 year warranty

More About this Item

Jar testers are designed to test the efficiency of flocculation or precipitation reagents on a small scale. Stirring rod shafts can individually be stopped and height can be adjusted while the system is operating. Choose from portable or laboratory testers.

Portable Jar Tester is available in a space saving 4-position model with stirring positions around a central lamp. Set stirring speed mechanically to 20, 40, 50, 100 or 120 rpm, and set time for 1 to 30 minutes. Central lamp makes it easier to observe flocculation process. Portable tester requires 1-L beakers (order separately, 34512-11 6/pk).

Two year warrantyConformite Europeenne
Specifications
  • Meter TypeBenchtop
  • Display typeAnalog
  • Power (VAC)100 to 240
  • Power (Hz)50 or 60
  • Width (cm)25.4
  • Height (cm)31.75
  • Width (in)10
  • Length (in)10
  • Height (in)12 1/2
  • Number of Positions4
  • Stirring Control20, 40, 50, 100, 120 RPM
  • Timer Control1 to 30 min
  • DescriptionFP4 Portable 4-position Flocculator 100-240V
  • Warranty2 year
Reviews
Q&A