Velp F30520201 FIWE3 Raw fiber extractor 230V/50Hz

Representative image only.

Velp F30520201 FIWE3 Raw fiber extractor 230V/50Hz

Velp | Mfr: F30520201| Item: 1595664

Rapid analysis in just two hours!

Call For Price

Call For Price
Product Description

Includes

heat shield, three glass crucibles, crucible holder, hot plate, glass reagent bottle, PVC tube, inlet tube, and pincers.

Key Features

  • Reliable results with high ±1% reproducibility
  • No sample loss-same crucibles used for weighing, drying, and washing
  • Complete system includes hot plate for preheating the reagents
  • 3 year warranty

More About this Item

Versatile instrument is suitable for the crude fiber analysis and detergent fiber extractions-such as total raw fiber, neutral and acid detergent, and fiber fraction analysis-according to official standard methods. This three-position unit performs single or multiple extraction including boiling, rinsing, and filtration. The extremely efficient heating element speeds up the analysis, saving you time.
The Weende method is one of the oldest analytical procedure in use today, in which fiber concentration is measured as crude fiber (CF). Weende method has been now largely replaced by the neutral detergent fiber (NDF), a method developed by Van Soest, being now the most common measure of fiber used for animal feed analysis.
The fiber analyzer is also suitable for Acid Detergent Lignin (ADL) determination and Wijkstrom technique, a modification of Weende method, and Acid Detergent Fiber (ADF).
Specifications:
Construction material: Epoxy painted stainless steel structure|Number of samples: 3
Digital timer: 0 - 99 minutes with acoustic signal
Type of extractions: Hot and cold
Sample removal: Air pump
Reagent discharge: Peristaltic pump
Temperature: Electronic regulation
Reagents and cooling water: Separated outlets
Sample quantity: From 0.5 to 3 g
Reproducibility: ± 1%

3 Year Warranty
Specifications
  • DescriptionFIWE3 Raw fiber extractor 230V/50Hz
  • Warranty3 year
Reviews
Q&A