Velp F208B0063 VTP Digital Temperature Controller
Representative image only.
Velp F208B0063 VTP Digital Temperature Controller
Velp | Mfr: F208B0063| Item: 8657936
Connects easily to hot plate stirrers for direct temperature control
₹60,360.00INR / Each
₹60,360.00INR / Each
Product Description
Includes
Temperature probe, support rod, clamps, and power cable.
Key Features
  • Incorporates timer for unsupervised operation and increased productivity
  • Ensures control accuracy of ± 0.5 °C
  • Direct connection to the hot plate stirrers
More About this Item
The digital temperature controller with a pre-set timer offers direct temperature control up to 572°F (300°C) with an accuracy of ±0.5°C. It features a LCD control panel for easy access and visibility. The controller is ready to use and can be combined with most common hot plate stirrers.
Two year warranty
Specifications
  • DescriptionAccessory Vertex Temperature Controller
  • Warranty2-1/2 Years