Velp F105A0108 JLT4 Laboratory 4-Position Flocculator 100-240V

Representative image only.

Velp F105A0108 JLT4 Laboratory 4-Position Flocculator 100-240V

Velp | Mfr: F105A0108| Item: 9956113

Save money and chemicals by testing on a small scale

₹341,310.00INR / Each

₹341,310.00INR / Each

Buy It Together

Current Item

Velp F105A0108 JLT4 Laboratory 4-Position Flocculator 100-240V
Velp F105A0108 JLT4 Laboratory 4-Position Flocculator 100-240V
9956113

₹341,310.00INR / Each
+ + =
Total Price 345410.00 ADD 3 ITEMS TO CART
Product Description

Includes

stainless steel stirring rods and universal AC adapter.

Key Features

  • ASTM D2035 compliant method
  • Single motor drives all positions at a consistent speed
  • Choose from portable or laboratory models, with four or six positions
  • 2 year warranty

More About this Item

Jar testers are designed to test the efficiency of flocculation or precipitation reagents on a small scale. Stirring rod shafts can individually be stopped and height can be adjusted while the system is operating. Choose from portable or laboratory testers.

Laboratory Jar Tester is available with four positions with a digital display that shows stirring speed and time. Stirring speed is programmable from 10 to 300 rpm, and time from 1 to 999 minutes or 0 to 99 hours. Featuring an illuminated back panel for enhanced visibility. Laboratory units require standard 1-L beakers (order separately, 34512-11).

Two year warrantyConformite Europeenne
Specifications
  • Meter TypeBenchtop
  • Display typeAnalog
  • Power (VAC)100 to 240
  • Power (Hz)50 or 60
  • Width (cm)64.77
  • Height (cm)34.29
  • Width (in)25 1/2
  • Length (in)10 1/4
  • Height (in)13 1/2
  • Number of Positions4
  • Stirring Control10 to 300 RPM
  • Timer Control1 to 999 min, 1 to99 hrs, Continuous
  • DescriptionJLT4 Laboratory 4-Position Flocculator 100-240V
  • Warranty2 year
Reviews
Q&A