Vanguard Safety R18G48 A4 HPPE Uncoated Cut Resistant Work Gloves, Gray, Medium; 24 Pairs/CS
Representative image only.
Vanguard Safety R18G48 A4 HPPE Uncoated Cut Resistant Work Gloves, Gray, Medium; 24 Pairs/CS
Vanguard Safety | Mfr: R18G48| Item: 7890718
Extended cuff protects wrists and forearms
₹40,100.00INR / Case of 48
₹40,100.00INR / Case of 48
Product Description
Key Features
  • Seamless knit HPPE blended liner keeps hands cool and dry
  • Ambidextrous
  • FFDCA compliant for food handling
  • EN 388 Standard with 3X41D rating
  • ANSI abrasion level 3
  • ANSI A4 cut level
  • Machine washable
More About this Item
These A4 uncoated work gloves provide excellent dexterity for use in the food processing, food service, and manufacturing environments. An extended cuff protects wrists and forearms. The gloves may be wore as a cut protection liner under disposable and chemical resistant gloves.
Specifications
  • SizeMedium
  • Length (in)9 29/64
  • Length (cm)24
  • ColorGray
  • Lining13g High Performance Polyethylene (HPPE)
  • CuffExtended
  • ComplianceFFDCA compliant for food handling
  • ApprovalANSI 3 ABR, ANSI A4 cut
  • SterileNo
  • PowderedNo
  • DescriptionA4 HPPE Uncoated Cut Resistant Gloves, Gray, Medium; 24 Pairs/CS