Vanguard Safety R11A36 A3 HPPE PU Coated Cut Resistant Work Gloves, Salt & Pepper, XS; 144 Pairs/CS
Representative image only.
Vanguard Safety R11A36 A3 HPPE PU Coated Cut Resistant Work Gloves, Salt & Pepper, XS; 144 Pairs/CS
Vanguard Safety | Mfr: R11A36| Item: 7890712
Palm coating resists abrasion, provides good wet and dry grip
₹201,000.00INR / Case of 288
₹201,000.00INR / Case of 288
Product Description
Key Features
  • Seamless knit HPPE blended liner keeps hands cool
  • FFDCA compliant for food handling
  • EN 388 Standard with 4X43C rating
  • ANSI abrasion level 5
  • ANSI A3 cut level
  • Machine washable
More About this Item
These A3 polyurethane palm coated work gloves are cut and abrasion resistant and offer excellent gripping while wet or dry. The breathable, seamless knit high performance polyethylene (HPPE) liner keeps your hands cool. The gloves are ideal for manufacturing, construction, metal and glass fabrication and machining work.
Specifications
  • SizeExtra small
  • Length (in)8 17/64
  • Length (cm)21
  • ColorGray
  • Lining13g High Performance Polyethylene (HPPE)
  • ComplianceFFDCA compliant for food handling
  • ApprovalANSI 5 ABR, ANSI A3 cut
  • SterileNo
  • PowderedNo
  • DescriptionA4 HPPE PU Coated Cut Resistant Gloves, Salt & Pepper, XS; 144 Pairs/CS