Vanguard Safety Powder-Free Vinyl Exam Gloves, 4 mil, Clear, Small; 1000/CS
Representative image only.
Vanguard Safety Powder-Free Vinyl Exam Gloves, 4 mil, Clear, Small; 1000/CS
Vanguard Safety | Mfr: A21A11| Item: 7890600
Economical choice for light duty maintenance
₹9,800.00INR / Case of 1000
₹9,800.00INR / Case of 1000
Product Description
Key Features
  • Powder-free
  • Medical grade – ASTM D6319
  • Prop 65 Compliant
  • Complies with FFDCA food handling requirements 21 CFR, Parts 170-199
  • Ambidextrous
  • 2 month warranty
More About this Item
These 4-mil thick clear disposable vinyl exam gloves with a smooth texture are ideal for light duty maintenance. Powder-free to reduce skin irritation. Applications include healthcare, dental, food service, food processing, manufacturing, and laboratory industries.
2 Month Warranty
Specifications
  • MaterialVinyl
  • Thickness (mil)4
  • SizeSmall
  • Length (in)8 45/64
  • Length (cm)22
  • ColorClear
  • ComplianceFDA approved
  • SterileNo
  • Palm Thickness (mil)3
  • PowderedNo
  • DescriptionPowder-Free Vinyl Exam Gloves, 4 mil, Clear, Small; 1000/CS
  • Warranty2 months