Vanguard Safety Powder-Free Nitrile Exam Gloves, 3.5 mil, Blue, Small; 1000/CS
Representative image only.
Vanguard Safety Powder-Free Nitrile Exam Gloves, 3.5 mil, Blue, Small; 1000/CS
Vanguard Safety | Mfr: A11A11| Item: 7890629
Protects against chemicals, oils, and blood borne pathogens
₹11,100.00INR / Case of 1000
₹11,100.00INR / Case of 1000
Product Description
Key Features
  • Texture fingertips for secure grip
  • Powder-free
  • Chemotherapy rated
  • Medical grade – ASTM D6319
  • Complies with FFDCA food handling requirements 21 CFR, Parts 170-199
  • Ambidextrous
  • 2 month warranty
More About this Item
The 3.5-mil thick nitrile gloves provide excellent puncture and abrasion resistance along with textured fingertips for a secure grip. The gloves are powder-free to reduce skin irritation and latex free. Ideal for use in chemotherapy, janitorial, dental, food service, healthcare, manufacturing, vet, and laboratory industries.
2 Month Warranty
Specifications
  • Thickness (mil)3.5
  • MaterialNitrile
  • SizeSmall
  • Length (in)8 45/64
  • Length (cm)22
  • ColorBlue
  • GripTextured
  • ComplianceFDA approved
  • SterileNo
  • Palm Thickness (mil)2.8
  • PowderedNo
  • DescriptionPowder-Free Nitrile Exam Gloves, 3.5 mil, Blue, Small; 1000/CS
  • Warranty2 months