Vanguard Safety Nitrile Unlined Gloves, 22 mil, 18" Cuff, Green, Extra-Large; 72/CS
Representative image only.
Vanguard Safety Nitrile Unlined Gloves, 22 mil, 18" Cuff, Green, Extra-Large; 72/CS
Vanguard Safety | Mfr: C16A210| Item: 7890675
Maintain secure grip in dry, wet, and oily conditions
₹113,000.00INR / Case of 72
₹113,000.00INR / Case of 72
Product Description
Key Features
  • Diamond textured non-slip surface ensures secure grip
  • Straight cuff
  • Complies with FFDCA food handling requirements 21 CFR, Parts 170-199
  • 2 month warranty
More About this Item
These reusable nitrile unlined gloves offer you durable protection from grease, fats, and heavy-duty cleaning solutions. Diamond textured non-slip surface offers secure grip in dry, wet, and oily conditions. The 22-mil thick gloves are puncture and abrasion resistant. Ideal for use in food processing and manufacturing plants.
2 Month Warranty
Specifications
  • Thickness (mil)22
  • MaterialNitrile
  • SizeExtra Large
  • ColorGreen
  • LiningUnlined
  • GripDiamond
  • Cuff18" Straight
  • ComplianceFFDCA compliant
  • SterileNo
  • Description Nitrile Unlined Gloves, 22 mil, 18" Cuff, Green, Extra-Large; 72/CS
  • Warranty2 months