Vanguard Safety Nitrile Flock Lined Gloves, 15 mil, 13" Cuff, Green, Extra-Large; 144/CS
Representative image only.
Vanguard Safety Nitrile Flock Lined Gloves, 15 mil, 13" Cuff, Green, Extra-Large; 144/CS
Vanguard Safety | Mfr: C14B210| Item: 7890671
Maintain secure grip in dry, wet, and oily conditions
₹46,100.00INR / Case of 144
₹46,100.00INR / Case of 144
Product Description
Key Features
  • Diamond textured non-slip surface ensures secure grip
  • Straight cuff
  • Complies with FFDCA food handling requirements 21 CFR, Parts 170-199
  • 2 month warranty
More About this Item
These reusable nitrile flock lined gloves offer you durable protection from grease, fats, and heavy-duty cleaning solutions. Diamond textured non-slip surface offers secure handling in dry, wet, and oily conditions. The 15-mil thick gloves are puncture and abrasion resistant. Ideal for use in food processing and manufacturing plants.
2 Month Warranty
Specifications
  • Thickness (mil)15
  • MaterialNitrile
  • SizeExtra Large
  • ColorGreen
  • LiningFlock
  • GripDiamond
  • Cuff13" Straight
  • ComplianceFFDCA compliant
  • SterileNo
  • DescriptionNitrile Flock Lined Gloves, 15 mil, 13" Cuff, Green, Extra-Large; 144/CS
  • Warranty2 months