Vanguard Safety G11C110 Flat Nitrile Coated Nylon Knit Gloves, Gray/Black, XL; 144 Pairs/CS
Representative image only.
Vanguard Safety G11C110 Flat Nitrile Coated Nylon Knit Gloves, Gray/Black, XL; 144 Pairs/CS
Vanguard Safety | Mfr: G11C110| Item: 7890690
Offers a secure grip in wet and oily environments
₹94,300.00INR / Case of 288
₹94,300.00INR / Case of 288
Product Description
Key Features
  • Seamless knit nylon liner keeps hands cool and dry
  • FFDCA compliant for food handling
  • EN 388 Standard with 4121X rating
  • ANSI abrasion level 4
  • Machine washable
More About this Item
These dexterous, form-fitting general-purpose gloves feature a nitrile palm coating which resists punctures and abrasions. Ultra tacky, smooth surface provides you with a secure nonslip grip and maximum holding power. The gloves are idea for package handling, warehousing, general assembly, machining, and maintenance work.
Specifications
  • SizeExtra large
  • Length (in)10
  • Length (cm)25.5
  • ColorGray/Black
  • Lining13g Nylon
  • ComplianceFFDCA compliant for food handling
  • ApprovalANSI 4 ABR
  • SterileNo
  • PowderedNo
  • DescriptionFlat Nitrile Coated Nylon Knit Work Gloves, Gray/Black, XL; 144 Pairs/CS