Value Plastics Sanitary Fitting, Straight Adapter, Polypropylene, 3/4" Mini Sanitary x Cap; 100/Pk
Representative image only.
Value Plastics Sanitary Fitting, Straight Adapter, Polypropylene, 3/4" Mini Sanitary x Cap; 100/Pk
Value Plastics | Mfr: SFMNC-6005| Item: 5011867
Seal sanitary ports with a secure and leak-proof connection
₹170,000.00INR / Pkg of 100
₹170,000.00INR / Pkg of 100
Product Description
Key Features
  • Ideal for capping off fluid transfer system
  • Sanitary fittings provide a secure connection where preventing contamination is crucial
  • Constructed of ADCF polypropylene
More About this Item
These Value Plastics Sanitary Fitting Caps are designed to easily seal sanitary ports. Sanitary fittings ensure a reliable connection while facilitating easy cleaning and preventing the buildup of bacteria or other contaminants. Sanitary end caps are often used in food and beverage, pharmaceutical, and biotechnology applications where maintaining cleanliness is crucial. Transparent polypropylene (PP) is suitable for Gamma radiation, ethylene oxide (EtO), E-beam, and autoclave sterilization.
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
Specifications
  • Fitting MaterialPolypropylene
  • Width (InchShort)63/64
  • Height (InchShort)51/64
  • Sterilization MethodGamma radiation, ethylene oxide (EtO), E-beam, and autoclave
  • Connection TypeCap
  • Number Of Ports1
  • Sanitary Clamp Size (Port 1) (InchShort)0.75
  • ColorWhite
  • Fitting Width (in)63/64
  • Regulatory ApprovalsFDA;Non-DEHP;USP;RoHS;ADCF;Non-Phthalate
  • DescriptionSanitary Fitting, Straight Adapter, Polypropylene, 3/4" Mini Sanitary x Cap; 100/Pk