Value Plastics Luer to Threaded Fitting, Straight Adapter, Polypropylene, Female Luer Thread Style with 5/16" Hex x 1/4-28 UNF(M) Bottom Sealing Thread; 25/Pk
Representative image only.
Value Plastics Luer to Threaded Fitting, Straight Adapter, Polypropylene, Female Luer Thread Style with 5/16" Hex x 1/4-28 UNF(M) Bottom Sealing Thread; 25/Pk
Value Plastics | Mfr: BSFTLL-6005| Item: 5012092
Luer fittings allow for a simple yet secure connection
₹5,500.00INR / Pkg of 25
₹5,500.00INR / Pkg of 25
Buy It Together
Current Item
Value Plastics Luer to Threaded Fitting, Straight Adapter, Polypropylene, Female Luer Thread Style with 5/16" Hex x 1/4-28 UNF(M) Bottom Sealing Thread; 25/Pk
Value Plastics Luer to Threaded Fitting, Straight Adapter, Polypropylene, Female Luer Thread Style with 5/16" Hex x 1/4-28 UNF(M) Bottom Sealing Thread; 25/Pk
5012092
₹5,500.00INR / Pkg of 25
+ + + =
Total Price 16200.00 ADD 4 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Quick and easy assembly and disassembly without the need for tools
  • Thread design provides a secure closure—preventing contamination
  • Conforms to ISO-594-1 and ISO 594-2 standards
  • Constructed of ADCF polypropylene
More About this Item
The Value Plastics Female Luer Thread Style to UNF Threaded Adapter Fittings are designed to ensure a tight and reliable connection while allowing for quick and easy assembly and disassembly without the need for tools. The thread feature is built into the female luer fitting that ensures the fittings remain firmly connected until intentionally disconnected. Adapters are available in various materials to accommodate various applications and compliance requirements.
Transparent polypropylene (PP) is suitable for Gamma radiation, ethylene oxide (EtO), E-beam, and autoclave sterilization.
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
Certificate of Compliance
Specifications
  • Fitting MaterialPolypropylene
  • Width (InchShort)5/16
  • Height (InchShort)25/32
  • Number Of Ports2
  • Thread Size NPT(M) (Port 2) (InchShort)1/4
  • Fitting Width (in)5/16
  • Regulatory ApprovalsNon-DEHP;RoHS;FDA;Non-Phthalate;USP;ADCF
  • Luer Connection Type (Port 1)Female Luer Thread Style with 5/16" Hex
  • Luer Connection Type (Port 2)1/4-28 UNF(M) Bottom Sealing Thread
  • DescriptionLuer to Threaded Fitting, Straight Adapter, Polypropylene, Female Luer Thread Style with 5/16" Hex x 1/4-28 UNF(M) Bottom Sealing Thread; 25/Pk