Value Plastics Luer to Threaded Fitting, Straight Adapter, Clear Polycarbonate, Female Luer Thread Style with 5/16" Hex x 1/4-28 UNF(M); 25/Pk
Representative image only.
Value Plastics Luer to Threaded Fitting, Straight Adapter, Clear Polycarbonate, Female Luer Thread Style with 5/16" Hex x 1/4-28 UNF(M); 25/Pk
Value Plastics | Mfr: SFTLL-9| Item: 5012086
Luer fittings allow for a simple yet secure connection
₹5,700.00INR / Pkg of 25
₹5,700.00INR / Pkg of 25
Buy It Together
Current Item
Value Plastics Luer to Threaded Fitting, Straight Adapter, Clear Polycarbonate, Female Luer Thread Style with 5/16" Hex x 1/4-28 UNF(M); 25/Pk
Value Plastics Luer to Threaded Fitting, Straight Adapter, Clear Polycarbonate, Female Luer Thread Style with 5/16" Hex x 1/4-28 UNF(M); 25/Pk
5012086
₹5,700.00INR / Pkg of 25
+ + + =
Total Price 16500.00 ADD 4 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Quick and easy assembly and disassembly without the need for tools
  • Thread design provides a secure closure—preventing contamination
  • Conforms to ISO-594-1 and ISO 594-2 standards
  • Constructed of high temperature resistant polycarbonate
More About this Item
The Value Plastics Female Luer Thread Style to UNF Threaded Adapter Fittings are designed to ensure a tight and reliable connection while allowing for quick and easy assembly and disassembly without the need for tools. The thread feature is built into the female luer fitting that ensures the fittings remain firmly connected until intentionally disconnected. Adapters are available in various materials to accommodate various applications and compliance requirements.
Clear polycarbonate (PC) is suitable for sterilization via ethylene oxide (EtO).
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
Certificate of Compliance
Specifications
  • Fitting MaterialPolycarbonate
  • Width (InchShort)5/16
  • Height (InchShort)19/32
  • Number Of Ports2
  • Thread Size NPT(M) (Port 2) (InchShort)1/4
  • Fitting Width (in)5/16
  • Regulatory ApprovalsNon-DEHP;RoHS;Non-Phthalate;FDA
  • Luer Connection Type (Port 1)Female Luer Thread Style with 5/16" Hex
  • Luer Connection Type (Port 2)1/4-28 UNF(M) Thread
  • DescriptionLuer to Threaded Fitting, Straight Adapter, Clear Polycarbonate, Female Luer Thread Style with 5/16" Hex x 1/4-28 UNF(M); 25/Pk