Value Plastics Luer to Threaded Fitting, Elbow, White Nylon, Female Luer Thread Style with 7/16" Hex x 1/8-27 NPT(M); 25/Pk
Representative image only.
Value Plastics Luer to Threaded Fitting, Elbow, White Nylon, Female Luer Thread Style with 7/16" Hex x 1/8-27 NPT(M); 25/Pk
Value Plastics | Mfr: 18LFTLL-1| Item: 5012071
Luer fittings allow for a simple yet secure connection
₹8,200.00INR / Pkg of 25
₹8,200.00INR / Pkg of 25
Product Description
Key Features
  • Quick and easy assembly and disassembly without the need for tools
  • Thread design provides a secure closure—preventing contamination
  • Constructed of durable chemical resistance nylon
  • Conforms to ISO-594-1 and ISO 594-2 standards
More About this Item
The Value Plastics Female Luer Thread Style to NPT Adapter Fittings are designed to ensure a tight and reliable connection while allowing for quick and easy assembly and disassembly without the need for tools. The thread feature is built into the female luer fitting that ensures the fittings remain firmly connected until intentionally disconnected. NPT (National Pipe Thread) is a type of tapered thread commonly used for sealing and connecting pipe fittings and components. NPT thread fittings are designed to create a tight seal when assembled, preventing leakage of fluids or gases under pressure. The elbow orientation offers directional flexibility, while the barbs grip the inside of the hose, creating a tight seal to prevent leaks or disconnection. White nylon is suitable for Gamma radiation, ethylene oxide (EtO), and autoclave sterilization.
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
Certificate of Compliance
Specifications
  • Fitting MaterialNylon
  • Width (InchShort)15/16
  • Height (InchShort)43/64
  • Number Of Ports2
  • Thread Size NPT(M) (Port 2) (InchShort)1/8
  • Fitting Width (in)15/16
  • Regulatory ApprovalsRoHS;Non-DEHP;Non-Phthalate
  • Luer Connection Type (Port 1)Female Luer Thread
  • Luer Connection Type (Port 2)1/8-27 NPT(M) Thread
  • DescriptionLuer to Threaded Fitting, Elbow, White Nylon, Female Luer Thread Style with 7/16" Hex x 1/8-27 NPT(M); 25/Pk