Value Plastics Luer to Hose Barb Fitting, Straight Adapter, White Nylon, Male Luer Integral Lock Ring x 500 Series Barb 1/4" ID; 25/Pk
Representative image only.
Value Plastics Luer to Hose Barb Fitting, Straight Adapter, White Nylon, Male Luer Integral Lock Ring x 500 Series Barb 1/4" ID; 25/Pk
Value Plastics | Mfr: MTLL055-1| Item: 5012019
Luer tapers that are constructed to meet ISO 594-1 and 594-2 standards
₹2,700.00INR / Pkg of 25
₹2,700.00INR / Pkg of 25
Product Description
Key Features
  • Rotating locks minimize twisting and kinking of tubing during positioning
  • Tapered cone seals are reliable and uncomplicated
  • Facilitate single-use configurations that minimize downtime
More About this Item
These luer fittings come in many configurations and several materials, joining tubing from 1/16” (1.6 mm) ID to 1/4” (6.4 mm) ID. The integral lock ring is a built-in mechanism that provides a secure connection to the female luer to prevent accidental disconnection and stop leakage or contamination. The 500 series barb is composed of blended radii giving it a distinct look. Each barb is designed to accommodate a range of tube sizes and tube types. Consideration should be given to flow restriction, pressure capability, and the force required for assembly of the tube. Because the fittings are made to ISO standards that are well understood globally, it ensures proper interconnections and often removes the need to retrofit*.
Options include nylon or polypropylene resin.
Common features of nylon include a high mechanical strength, great balance of stiffness and toughness, good performance at high temperatures, good electrical and flammability properties, and resistance against abrasion or chemicals. It can be sterilized using gamma radiation, Ethylene oxide (EtO), or autoclave.
Polypropylene is clarified and produced without any animal derived components or phthalates. It can be sterilized using gamma radiation, Ethylene oxide (EtO), E-beam, or autoclave.
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
Certificate of Compliance
Specifications
  • Fitting MaterialNylon
  • Number Of Ports2
  • Barb Size / Tubing Id (Port 2) (in)1/4
  • Regulatory ApprovalsRoHS;Non-DEHP;Non-Phthalate
  • Luer Connection Type (Port 1)Male Luer
  • Luer Connection Type (Port 2)Hose Barb
  • DescriptionLuer to Hose Barb Fitting, Straight Adapter, White Nylon, Male Luer Integral Lock Ring x 500 Series Barb 1/4" ID; 25/Pk