Value Plastics Luer to Hose Barb Fitting, Straight Adapter, Polypropylene, Male Luer Integral Lock Ring x 300 Series Barb 1/4" ID; 25/Pk
Representative image only.
Value Plastics Luer to Hose Barb Fitting, Straight Adapter, Polypropylene, Male Luer Integral Lock Ring x 300 Series Barb 1/4" ID; 25/Pk
Value Plastics | Mfr: MTLL360-6005| Item: 5012033
Luer tapers that are constructed to meet ISO 594-1 and 594-2 standards
₹3,900.00INR / Pkg of 25
₹3,900.00INR / Pkg of 25
Product Description
Key Features
  • Rotating locks minimize twisting and kinking of tubing during positioning
  • Tapered cone seals are reliable and uncomplicated
  • Facilitate single-use configurations that minimize downtime
More About this Item
These luer fittings come in many configurations and several materials, joining tubing from 1/16” (1.6 mm) ID to 1/4” (6.4 mm) ID. The integral lock ring is a built-in mechanism that provides a secure connection to the female luer to prevent accidental disconnection and stop leakage or contamination. The 300 series barb offers outstanding performance characteristics when used with flexible tubing. Tubing is expanded by 35% above its nominal inside diameter, permitting high-pressure capabilities in the larger tube sizes. This barb style provides high flow efficiency and excellent tensile strength. Because the fittings are made to ISO standards that are well understood globally, it ensures proper interconnections and often removes the need to retrofit*.
Polypropylene is clarified and produced without any animal derived components or phthalates. It can be sterilized using gamma radiation, Ethylene oxide (EtO), E-beam, or autoclave.
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
Certificate of Compliance
Specifications
  • Fitting MaterialPolypropylene
  • Number Of Ports2
  • Barb Size / Tubing Id (Port 2) (in)1/4
  • Regulatory ApprovalsNon-DEHP;FDA;RoHS;USP;Non-Phthalate;ADCF
  • Luer Connection Type (Port 2)Hose Barb
  • DescriptionLuer to Hose Barb Fitting, Straight Adapter, Polypropylene, Male Luer Integral Lock Ring x 300 Series Barb 1/4" ID; 25/Pk