Value Plastics Luer Fitting, White Nylon, Male Luer Integral Lock Ring x Female Luer Thread; 25/Pk
Representative image only.
Value Plastics Luer Fitting, White Nylon, Male Luer Integral Lock Ring x Female Luer Thread; 25/Pk
Value Plastics | Mfr: LC34-1| Item: 5012003
Prevent leaks with integral lock ring mechanism
₹3,100.00INR / Pkg of 25
₹3,100.00INR / Pkg of 25
Buy It Together
Current Item
Value Plastics Luer Fitting, White Nylon, Male Luer Integral Lock Ring x Female Luer Thread; 25/Pk
Value Plastics Luer Fitting, White Nylon, Male Luer Integral Lock Ring x Female Luer Thread; 25/Pk
5012003
₹3,100.00INR / Pkg of 25
+
Cole-Parmer TPV Industrial Grade Tubing, 1/4" ID x 1/2" OD; 50 Ft
5011587
₹35,900.00INR / Pkg of 1
+ =
Total Price 40400.00 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Luer fittings allow for a simple yet secure twist-lock connection
  • Easily transition from a male luer to female luer fitting
  • Ideal for general laboratory and single-use applications
  • Constructed of durable chemical resistance nylon
More About this Item
Value Plastics Male Luer Integral Lock Ring to Female Luer Thread Style Couplers are designed to easily transition your fluid transfer system from a male to female luer connection. Luer fittings ensure a tight and reliable connection while allowing for quick and easy assembly and disassembly without the need for tools. The integral lock ring is a feature built into the male luer fitting that ensures the fittings remain firmly connected until intentionally disconnected. These adapters are available in various materials to accommodate various applications and compliance requirements.
White nylon is suitable for Gamma radiation, ethylene oxide (EtO), and autoclave sterilization.
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
Certificate of Compliance
Specifications
  • Fitting MaterialNylon
  • Width (InchShort)7/16
  • Height (InchShort)43/64
  • Number Of Ports2
  • Fitting Width (in)7/16
  • Regulatory ApprovalsNon-DEHP;RoHS;Non-Phthalate
  • Luer Connection Type (Port 1)Male Luer Integral Lock Ring
  • Luer Connection Type (Port 2)Female Luer Thread
  • DescriptionLuer Fitting, White Nylon, Male Luer Integral Lock Ring x Female Luer Thread; 25/Pk