Value Plastics Luer Fitting, Union, White Nylon, Male Luer Slip; 25/Pk
Representative image only.
Value Plastics Luer Fitting, Union, White Nylon, Male Luer Slip; 25/Pk
Value Plastics | Mfr: MTLCS-1| Item: 5012014
Luer fittings allow for a simple yet secure connection
₹2,800.00INR / Pkg of 25
₹2,800.00INR / Pkg of 25
Buy It Together
Current Item
Value Plastics Luer Fitting, Union, White Nylon, Male Luer Slip; 25/Pk
Value Plastics Luer Fitting, Union, White Nylon, Male Luer Slip; 25/Pk
5012014
₹2,800.00INR / Pkg of 25
+
Cole-Parmer TPV Industrial Grade Tubing, 1/4" ID x 1/2" OD; 50 Ft
5011587
₹35,900.00INR / Pkg of 1
=
Total Price 38700.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Quick and easy assembly and disassembly without the need for tools
  • Constructed of durable chemical resistance nylon
  • Conforms to ISO-594-1 and ISO 594-2 standards
More About this Item
The Male Luer Slip Union Fittings are designed to ensure a tight and reliable connection while allowing for quick and easy assembly and disassembly without the need for tools. White nylon is suitable for Gamma radiation, ethylene oxide (EtO), and autoclave sterilization.
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
Certificate of Compliance
Specifications
  • Fitting MaterialNylon
  • Width (InchShort)3/8
  • Height (InchShort)15/16
  • Number Of Ports2
  • Fitting Width (in)3/8
  • Regulatory ApprovalsNon-Phthalate;RoHS;Non-DEHP
  • Luer Connection Type (Port 1)Male Luer Slip
  • Luer Connection Type (Port 2)Male Luer Slip
  • DescriptionLuer Fitting, Union, White Nylon, Male Luer Slip; 25/Pk