Value Plastics Luer Fitting, Tee Union, White Nylon, Female Luer; 25/Pk
Representative image only.
Value Plastics Luer Fitting, Tee Union, White Nylon, Female Luer; 25/Pk
Value Plastics | Mfr: FTLT-1| Item: 5011997
Easily transition to a male luer fitting with directional flexibility
₹5,200.00INR / Pkg of 25
₹5,200.00INR / Pkg of 25
Product Description
Key Features
  • Quick and easy assembly and disassembly without the need for tools
  • Luer fittings allow for a simple yet secure connection
  • Conforms to ISO-594-1 and ISO 594-2 standards
  • Constructed of durable chemical resistance nylon
More About this Item
These Value Plastics Female Luer Tee Fittings are designed to ensure a tight and reliable connection while allowing for quick and easy assembly and disassembly without the need for tools. The tee orientation offers directional flexibility, creating a tight seal to prevent leaks or disconnection. These adapters are available in various materials to accommodate various applications and compliance requirements.
White nylon is suitable for Gamma radiation, ethylene oxide (EtO), and autoclave sterilization.
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
Certificate of Compliance
Specifications
  • Fitting MaterialNylon
  • Width (InchShort)5/8
  • Height (InchShort)15/16
  • Number Of Ports3
  • Fitting Width (in)5/8
  • Regulatory ApprovalsNon-DEHP;Non-Phthalate;RoHS
  • Luer Connection Type (Port 1)Female Luer
  • Luer Connection Type (Port 2)Female Luer
  • Luer Connection Type (Port 3)Female Luer
  • DescriptionLuer Fitting, Tee Union, White Nylon, Female Luer; 25/Pk