Value Plastics Luer Fitting, Tee Union, Clear Polycarbonate, Female Luer Lug; 25/Pk
Representative image only.
Value Plastics Luer Fitting, Tee Union, Clear Polycarbonate, Female Luer Lug; 25/Pk
Value Plastics | Mfr: FTLT-9| Item: 5011999
Easily transition to a male luer fitting with directional flexibility
₹6,400.00INR / Pkg of 25
₹6,400.00INR / Pkg of 25
Product Description
Key Features
  • Quick and easy assembly and disassembly without the need for tools
  • Luer fittings allow for a simple yet secure connection
  • Conforms to ISO-594-1 and ISO 594-2 standards
  • Constructed of high temperature resistant polycarbonate
More About this Item
These Value Plastics Female Luer Tee Fittings are designed to ensure a tight and reliable connection while allowing for quick and easy assembly and disassembly without the need for tools. The tee orientation offers directional flexibility, creating a tight seal to prevent leaks or disconnection. These adapters are available in various materials to accommodate various applications and compliance requirements.
Clear polycarbonate (PC) is suitable for sterilization via ethylene oxide (EtO).
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
Certificate of Compliance
Specifications
  • Fitting MaterialPolycarbonate
  • Width (InchShort)5/8
  • Height (InchShort)15/16
  • Number Of Ports3
  • Fitting Width (in)5/8
  • Regulatory ApprovalsNon-Phthalate;FDA;Non-DEHP;RoHS
  • Luer Connection Type (Port 1)Female Luer
  • Luer Connection Type (Port 2)Female Luer
  • Luer Connection Type (Port 3)Female Luer
  • DescriptionLuer Fitting, Tee Union, Clear Polycarbonate, Female Luer Lug; 25/Pk