Value Plastics Luer Fitting, Tee Adapter, Polypropylene, Male Luer Slip x Male Luer Slip x Female Luer Thread; 25/Pk
Representative image only.
Value Plastics Luer Fitting, Tee Adapter, Polypropylene, Male Luer Slip x Male Luer Slip x Female Luer Thread; 25/Pk
Value Plastics | Mfr: LT787-6005| Item: 5012010
Easily transition to a luer fitting with directional flexibility
₹8,800.00INR / Pkg of 25
₹8,800.00INR / Pkg of 25
Product Description
Key Features
  • Quick and easy assembly and disassembly without the need for tools
  • Luer fittings allow for a simple yet secure connection
  • Conforms to ISO-594-1 and ISO 594-2 standards
  • Constructed of ADCF polypropylene
More About this Item
These Value Plastics Luer Elbow Fittings are designed to ensure a tight and reliable connection while allowing for quick and easy assembly and disassembly without the need for tools. The tee orientation offers directional flexibility, creating a tight seal to prevent leaks or disconnection. Transparent polypropylene (PP) is suitable for Gamma radiation, ethylene oxide (EtO), E-beam, and autoclave sterilization.
May be used with separate rotating lock ring, 50115-54, sold separately.
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
Certificate of Compliance
Specifications
  • Fitting MaterialPolypropylene
  • Width (InchShort)29/32
  • Height (InchShort)1 7/32
  • Connection TypeMale Luer Slip x Male Luer Slip x Female Luer Thread
  • Number Of Ports3
  • Flow PatternTee
  • Fitting Width (in)29/32
  • Regulatory ApprovalsFDA;RoHS;USP;Non-DEHP;Non-Phthalate;ADCF
  • Luer Connection Type (Port 1)Female Luer Thread
  • Luer Connection Type (Port 2)Male Slip Luer
  • DescriptionLuer Fitting, Tee Adapter, Polypropylene, Male Luer Slip x Male Luer Slip x Female Luer Thread; 25/Pk