Value Plastics Luer Fitting, Tee Adapter, Polypropylene, Female Luer Thread x Female Luer Thread x Male Luer Slip; 25/Pk
Representative image only.
Value Plastics Luer Fitting, Tee Adapter, Polypropylene, Female Luer Thread x Female Luer Thread x Male Luer Slip; 25/Pk
Value Plastics | Mfr: LT878-6005| Item: 5012011
Easily transition to a luer fitting with directional flexibility
₹8,800.00INR / Pkg of 25
₹8,800.00INR / Pkg of 25
Product Description
Key Features
  • Quick and easy assembly and disassembly without the need for tools
  • Luer fittings allow for a simple yet secure connection
  • Conforms to ISO-594-1 and ISO 594-2 standards
  • Constructed of ADCF polypropylene
More About this Item
These Value Plastics Luer Elbow Fittings are designed to ensure a tight and reliable connection while allowing for quick and easy assembly and disassembly without the need for tools. The tee orientation offers directional flexibility, creating a tight seal to prevent leaks or disconnection. Transparent polypropylene (PP) is suitable for Gamma radiation, ethylene oxide (EtO), E-beam, and autoclave sterilization.
May be used with separate rotating lock ring, 50115-54, sold separately.
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
Certificate of Compliance
Specifications
  • Fitting MaterialPolypropylene
  • Width (InchShort)37/64
  • Height (InchShort)1 7/32
  • Number Of Ports3
  • Fitting Width (in)37/64
  • Regulatory ApprovalsNon-DEHP;Non-Phthalate;RoHS;ADCF;FDA;USP
  • Luer Connection Type (Port 1)Female Luer Thread
  • Luer Connection Type (Port 2)Female Luer Thread
  • Luer Connection Type (Port 3)Male Slip Luer
  • DescriptionLuer Fitting, Tee Adapter, Polypropylene, Female Luer Thread x Female Luer Thread x Male Luer Slip; 25/Pk