Value Plastics Luer Fitting, Straight Adapter, Clear Polycarbonate, Male Luer Slip x Female Luer Thread; 25/Pk
Representative image only.
Value Plastics Luer Fitting, Straight Adapter, Clear Polycarbonate, Male Luer Slip x Female Luer Thread; 25/Pk
Value Plastics | Mfr: LC78-9| Item: 5012006
Luer fittings allow for a simple yet secure connection
₹3,200.00INR / Pkg of 25
₹3,200.00INR / Pkg of 25
Product Description
Key Features
  • Quick and easy assembly and disassembly without the need for tools
  • Thread design provides a secure closure—preventing contamination
  • Conforms to ISO-594-1 and ISO 594-2 standards
  • Constructed of high temperature resistant polycarbonate
More About this Item
The Male Luer Slip to Female Luer Thread Adapter Fittings are designed to ensure a tight and reliable connection while allowing for quick and easy assembly and disassembly without the need for tools. The thread feature is built into the female luer fitting that ensures the fittings remain firmly connected until intentionally disconnected. Adapters are available in various materials to accommodate various applications and compliance requirements.
Clear polycarbonate (PC) is suitable for sterilization via ethylene oxide (EtO).
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
Certificate of Compliance
Specifications
  • Fitting MaterialPolycarbonate
  • Width (InchShort)1/4
  • Height (InchShort)27/32
  • Number Of Ports2
  • Fitting Width (in)1/4
  • Regulatory ApprovalsNon-DEHP;RoHS;FDA;Non-Phthalate
  • Luer Connection Type (Port 1)Male Luer Slip
  • Luer Connection Type (Port 2)Female Luer Thread
  • DescriptionLuer Fitting, Straight Adapter, Clear Polycarbonate, Male Luer Slip x Female Luer Thread; 25/Pk