Value Plastics Luer Fitting, Finger Snap Luer Lock Ring, Clear Polycarbonate; 25/Pk
Representative image only.
Value Plastics Luer Fitting, Finger Snap Luer Lock Ring, Clear Polycarbonate; 25/Pk
Value Plastics | Mfr: FSLLR-9| Item: 5012058
Easily convert a male luer to male luer lock for enhanced leak prevention
₹2,400.00INR / Pkg of 25
₹2,400.00INR / Pkg of 25
Product Description
Key Features
  • Luer fittings allow for a simple yet secure twist-lock connection
  • Constructed of FDA compliant clear polycarbonate
  • Prevent leaks with lock ring mechanism
More About this Item
Value Plastics Value Plastics Finger Snap Luer Lock Rings are designed to easily transition your luer from a slip to locked connection. Luer fittings ensure a tight and reliable connection while allowing for quick and easy assembly and disassembly without the need for tools. This lock ring ensures the fittings remain firmly connected until intentionally disconnected. The finger snap design allows for easy manipulation and handling of the luer lock ring. Clear polycarbonate (PC) is suitable for sterilization via ethylene oxide (EtO).
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
Certificate of Compliance
Specifications
  • Fitting MaterialPolycarbonate
  • Width (InchShort)7/16
  • Height (InchShort)27/64
  • Fitting Width (in)7/16
  • Regulatory ApprovalsFDA;Non-DEHP;Non-Phthalate;RoHS
  • Luer Connection Type (Port 1)Finger Snap Luer Lock Ring
  • DescriptionLuer Fitting, Finger Snap Luer Lock Ring, Clear Polycarbonate; 25/Pk