Value Plastics Luer Fitting, Elbow Adapter, White Nylon, Male Luer x Female Luer Thread; 25/Pk
Representative image only.
Value Plastics Luer Fitting, Elbow Adapter, White Nylon, Male Luer x Female Luer Thread; 25/Pk
Value Plastics | Mfr: LE87-1| Item: 5012007
Easily transition to a luer fitting with directional flexibility
₹5,900.00INR / Pkg of 25
₹5,900.00INR / Pkg of 25
Buy It Together
Current Item
Value Plastics Luer Fitting, Elbow Adapter, White Nylon, Male Luer x Female Luer Thread; 25/Pk
Value Plastics Luer Fitting, Elbow Adapter, White Nylon, Male Luer x Female Luer Thread; 25/Pk
5012007
₹5,900.00INR / Pkg of 25
+
Cole-Parmer TPV Industrial Grade Tubing, 1/4" ID x 1/2" OD; 50 Ft
5011587
₹35,900.00INR / Pkg of 1
=
Total Price 41800.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Quick and easy assembly and disassembly without the need for tools
  • Luer fittings allow for a simple yet secure connection
  • Conforms to ISO-594-1 and ISO 594-2 standards
  • Constructed of durable chemical resistance nylon
More About this Item
These Value Plastics Luer Thread Elbow Fittings are designed to ensure a tight and reliable connection while allowing for quick and easy assembly and disassembly without the need for tools. The elbow orientation offers directional flexibility, creating a tight seal to prevent leaks or disconnection. These adapters are available in various materials to accommodate various applications and compliance requirements.
White nylon is suitable for Gamma radiation, ethylene oxide (EtO), and autoclave sterilization.
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
Certificate of Compliance
Specifications
  • Fitting MaterialNylon
  • Width (InchShort)37/64
  • Height (InchShort)29/32
  • Number Of Ports2
  • Fitting Width (in)37/64
  • Regulatory ApprovalsNon-Phthalate;Non-DEHP;RoHS
  • Luer Connection Type (Port 1)Male Luer
  • Luer Connection Type (Port 2)Female Luer Thread
  • DescriptionLuer Fitting, Elbow Adapter, White Nylon, Male Luer x Female Luer Thread; 25/Pk