Value Plastics Hose Barb Fitting, Tee Union, Polypropylene, 200 Series Barb, 1/8" ID; 25/Pk
Representative image only.
Value Plastics Hose Barb Fitting, Tee Union, Polypropylene, 200 Series Barb, 1/8" ID; 25/Pk
Value Plastics | Mfr: T230-6005| Item: 5012094
A secure connection under pressure with 200 series barbs
₹5,000.00INR / Pkg of 25
₹5,000.00INR / Pkg of 25
Product Description
Key Features
  • Quick and easy assembly and disassembly without the need for tools
  • Barbed ridges provide secure grip on the inside of flexible tubing
  • Tee orientation provides flexibility and versatility
  • Constructed of ADCF polypropylene
More About this Item
The Value Plastics 200 Series Tee Hose Barb Fittings are designed to provide outstanding performance characteristics when used with flexible tubing. Tubing is expanded by 50% above its nominal inside diameter, permitting high-pressure capabilities. The tee orientation offers directional flexibility, while the barbs grip the inside of the hose, creating a tight seal to prevent leaks or disconnection. These adapters are available in various materials to accommodate various applications and compliance requirements.
Transparent polypropylene (PP) is suitable for Gamma radiation, ethylene oxide (EtO), E-beam, and autoclave sterilization.
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
Certificate of Compliance
Specifications
  • Fitting MaterialPolypropylene
  • Sterilization MethodGamma radiation, ethylene oxide (EtO), E-beam, and autoclave
  • Number Of Ports3
  • Connection TypeHose Barb Union
  • Barb Size / Tubing Id (Port 1) (mm)3.175
  • Barb Size / Tubing Id (Port 1) (in)1/8
  • Barb Size / Tubing Id (Port 2) (mm)3.175
  • Barb Size / Tubing Id (Port 2) (in)1/8
  • Barb Size / Tubing Id (Port 3) (mm)3.175
  • Barb Size / Tubing Id (Port 3) (in)1/8
  • Flow PatternTee
  • ColorTransparent
  • Regulatory ApprovalsFDA;RoHS;USP;ADCF;Non-DEHP;Non-Phthalate
  • DescriptionHose Barb Fitting, Tee Union, Polypropylene, 200 Series Barb, 1/8" ID; 25/Pk