Value Plastics Hose Barb Fitting, Straight Reducer, Polypropylene, 500 Series Barb, 1/4" ID x 1/8" ID; 25/Pk
Representative image only.
Value Plastics Hose Barb Fitting, Straight Reducer, Polypropylene, 500 Series Barb, 1/4" ID x 1/8" ID; 25/Pk
Value Plastics | Mfr: N055/013-6005| Item: 5012093
A secure tubing connection with semi-rigid tubing
₹4,500.00INR / Pkg of 25
₹4,500.00INR / Pkg of 25
Product Description
Key Features
  • Quick and easy assembly and disassembly without the need for tools
  • Barbed ridges provide secure grip on the inside of flexible tubing
  • Constructed of USP compliant ADCF polypropylene
More About this Item
The Value Plastics 500 Series Hose Barb Reducer Fittings are designed with profiles composed of blended radii, giving this series a distinct look. Each barb is designed to accommodate a range of tube sizes and tube types. This series is ideal for semi-rigid tubing such as polyurethane and EVA. Transparent polypropylene (PP) is suitable for Gamma radiation, ethylene oxide (EtO), E-beam, and autoclave sterilization.
Constructed of ADCF polypropylene
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
Certificate of Compliance
Specifications
  • Fitting MaterialPolypropylene
  • Sterilization MethodGamma irradiation between 25 and 50 kGy, EtO, or autoclave up to 121°C (250°F)
  • Connection TypeHose Barb Reducer
  • Number Of Ports2
  • Barb Size / Tubing Id (Port 1) (mm)6.35
  • Barb Size / Tubing Id (Port 1) (in)1/4
  • Barb Size / Tubing Id (Port 2) (mm)3.175
  • Barb Size / Tubing Id (Port 2) (in)1/8
  • Flow PatternStraight
  • ColorTransparent
  • Regulatory ApprovalsADCF;FDA;USP;Non-Phthalate;RoHS;Non-DEHP
  • DescriptionHose Barb Fitting, Straight Reducer, Polypropylene, 500 Series Barb, 1/4" ID x 1/8" ID; 25/Pk