Value Plastics Hose Barb Fitting, Elbow Union, PVDF, 200 Series 1/8" ID; 25/Pk
Representative image only.
Value Plastics Hose Barb Fitting, Elbow Union, PVDF, 200 Series 1/8" ID; 25/Pk
Value Plastics | Mfr: L230-J2A| Item: 5012002
A secure connection under pressure with 200 series barbs
₹7,100.00INR / Pkg of 25
₹7,100.00INR / Pkg of 25
Product Description
Key Features
  • Quick and easy assembly and disassembly without the need for tools
  • Barbed ridges provide secure grip on the inside of flexible tubing
  • Constructed of USP compliant PVDF
More About this Item
The Value Plastics 200 Series Elbow Hose Barb Fittings are designed to provide outstanding performance characteristics when used with flexible tubing. Tubing is expanded by 50% above its nominal inside diameter, permitting high-pressure capabilities. The elbow orientation offers directional flexibility, while the barbs grip the inside of the hose, creating a tight seal to prevent leaks or disconnection. Transparent PVDF is suitable for Gamma radiation, ethylene oxide (EtO), E-beam, and autoclave sterilization.
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
Certificate of Compliance
Specifications
  • Fitting MaterialPVDF
  • Sterilization MethodGamma radiation, ethylene oxide (EtO), E-beam, and autoclave
  • Number Of Ports2
  • Connection TypeHose Barb Union
  • Barb Size / Tubing Id (Port 1) (mm)3.175
  • Barb Size / Tubing Id (Port 1) (in)1/8
  • Barb Size / Tubing Id (Port 2) (mm)3.175
  • Barb Size / Tubing Id (Port 2) (in)1/8
  • Flow PatternElbow
  • ColorTransparent
  • Regulatory ApprovalsFDA;Non-Phthalate;RoHS;ADCF;Non-DEHP;USP
  • DescriptionHose Barb Fitting, Elbow Union, PVDF, 200 Series 1/8" ID; 25/Pk