Vaisala Intrinsically Safe RH/Temperature Transmitter for High Humidity, 0 to 100% RH, 0 to 1 bar, –94 to 356°F, 2-m Cable

Representative image only.

Vaisala Intrinsically Safe RH/Temperature Transmitter for High Humidity, 0 to 100% RH, 0 to 1 bar, –94 to 356°F, 2-m Cable

Vaisala | Mfr: HMT370EX 75P2B2AE4DN| Item: 3780536

Safely measure humidity even in the most hazardous environments

₹2,141,000.00INR / Each

₹2,141,000.00INR / Each
Product Description

Includes

Traceable calibration certificate, two stahl barriers, 2-m cable, and 1/2" NPT conduit bushing.

Key Features

  • Rated intrinsically safe for operation in up to Zone 0/20
  • Detachable probe module—easily replace and remove for calibration
  • Vaisala HUMICAP® sensor provides quality and reliability
  • Configure and make adjustments on the graphical LCD display
  • Two 4 to 20 mA outputs
  • Additional monitoring and configuration options available with Vaisala Insight software
  • Constructed for installation in high humidity environments
  • 2 year warranty

More About this Item

Measure relative humidity and temperature in even the most hazardous area with the Vaisala intrinsically safe transmitters. Transmitters withstand continuous exposure to potentially explosive environments that contain flammable liquids, gases, or dust—rated for use in up to Zone 0 and 20.

Easily configure outputs and adjust measurement calibration directly on the local graphical display. Two 4 to 20 mA outputs provide connectivity options to automation systems. Connect the transmitter to free Vaisala Insight software with a USB cable (order separately) for additional configuration and monitoring options.

All probes feature a thin-film capacitive Vaisala HUMICAP sensor that offers excellent accuracy, long-term stability, and negligible hysteresis. Each probe comes with a detachable probe module allowing the probe to be easily calibrated outside the hazardous area without removing the entire transmitter. Select a transmitter with a probe option best suited for your application.

Two year warranty
Specifications
  • Min RH Range (%)0
  • Max RH Range (%)100
  • RH Accuracy±0.8% RH from 0 to 90% RH at 23°C (73.4°F)
  • Min Probe Temperature (DegreeF)-94
  • Min Probe Temperature (DegreeC)-70
  • Max Probe Temperature (DegreeF)356
  • Max Probe Temperature (DegreeC)180
  • Probe Temperature Accuracy±0.1°C (±0.18°F) at 23°C (73.4°F)
  • Response TimeRH: 15 s
  • Display TypeGraphical LCD
  • OutputTwo loop-powered 4 to 20 mA
  • Power (VDC)12 to 28
  • Length (in)3 15/16
  • Diameter (in)1/2
  • DescriptionIntrinsically Safe RH/Temperature Transmitter for High Humidity, 0 to 100% RH, 0 to 1 bar, –94 to 356°F, 2-m Cable
  • Warranty2 years
Reviews
Q&A