Vaisala HMT370 Wall-Mount Humidity Transmitter With Display

Representative image only.

Vaisala HMT370 Wall-Mount Humidity Transmitter With Display

Vaisala | Mfr: HMT370EX 15A1NB2AC1AN| Item: 3751012

Factory Mutual approved for hazardous indoor applications

Call For Price

for an additional
₹40,300.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services

Call For Price

Buy It Together

Current Item

Vaisala HMT370 Wall-Mount Humidity Transmitter With Display
Vaisala HMT370 Wall-Mount Humidity Transmitter With Display
3751012

Call For Price
+ =
Total Price 40300.00 ADD 2 ITEMS TO CART
Product Description

Key Features

  • Meets Class I, II, III, Division 1, Groups A-G and Division 2, Groups A-D, F and G
  • 1 year warranty

More About this Item

Measure temperature and humidity in environmentally hazardous areas. NEMA 4 (IP65) housing is rated water- and dust-proof. Wall-mount and duct-mount versions are available with or without an LCD. Two 4 to 20 mA outputs offer analog signals, one for humidity and one for temperature.
What’s included: certificate of NIST traceability. Wall-mount transmitters include a probe measuring 4 5/8”L x 1/2” dia. Duct-mount transmitters include a probe measuring 2 11/16”L x 1/2” dia with 78" (2 m) of cable length, and a duct installation kit.

IP 65NIST Traceable Calibration1 Year Warranty
Specifications
  • Min RH Range (%)0
  • Max RH Range (%)100
  • RH Accuracy±1% RH; ±2% RH above 90% RH
  • Min Temperature (° F)-40
  • Min Temperature (° C)-40
  • Max Temperature (° F)140
  • Max Temperature (° C)60
  • Temperature Accuracy±0.36°F (±0.2°C)
  • Response Time15 seconds
  • Display TypeLCD
  • Mounting TypeWall
  • Output4-20mA
  • Power (VDC)12 to 28
  • Height (in)4 1/2
  • Width (in)6 1/2
  • Depth (in)2 3/8
  • DescriptionWall-Mount Humidity Transmitter With Display
  • Warranty1 year
Reviews
Q&A