Vacuum Research 902130 Thermocouple Switch/transmitter, 1 To 1000 Mtorr
Representative image only.
Vacuum Research 902130 Thermocouple Switch/transmitter, 1 To 1000 Mtorr
Vacuum Research | Mfr: 902130| Item: 6880850
Use with Hastings® DV-6 or VRC gauge tubes
₹95,600.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
₹95,600.00INR / Each
Buy It Together
Current Item
Vacuum Research 902130 Thermocouple Switch/transmitter, 1 To 1000 Mtorr
Vacuum Research 902130 Thermocouple Switch/transmitter, 1 To 1000 Mtorr
6880850
₹95,600.00INR / Each
+ +
Vacuum Research 912086 Thermocouple Sensor, 1/8" NPT (m)
6880818
₹25,100.00INR / Each
=
Total Price 151400.00 ADD 3 ITEMS TO CART
Product Description
Key Features
  • Ranges from 10-5 Torr to 2 atmospheres
  • Analog output for PLC or computer interface
  • 2 year warranty
More About this Item
These active vacuum switch/transmitters provide an inexpensive and straightforward way to control your vacuum system. Use the Pirani sensor switch directly to initiate crossover after roughing, or the diaphragm sensor switch to control backfill pressure on pump and purge operations. Feed the switch output into your PC or PLC and let the digital system make the control decisions based on the output signal.To create a complete vacuum system, choose your vacuum range and sensor type from the table below, and then choose your sensor connection style. Switch/transmitter plugs directly into sensors. Measure high, medium, and low vacuum with Pirani, thermocouple, and diaphragm sensors.
Conformite EuropeenneTwo year warranty
Specifications
  • Max Vacuum (" Hg)39
  • Wetted Materials300 SS
  • Unit Of MeasurementmTorr
  • Battery Type12 VDC, 200 mA
  • Output0 to 2 VDC
  • DescriptionThermocouple Switch/transmitter, 1 To 1000 Mtorr