Utility Brush with 8" Handle, Nylon Bristles, White

Representative image only.

Utility Brush with 8" Handle, Nylon Bristles, White

| Item: 8455030

Effective cleaning for a variety of applications

₹2,400.00INR / Each

₹2,400.00INR / Each
Product Description

Key Features

  • Synthetic block is moisture and bacteria proof
  • Hang-up hole provide easy storage
  • Gnarled grip offers safe handling
  • 3 month warranty

More About this Item

The utility brushes come with medium-stif crimped bristles to help carry more water or solution to the cleaning surface. Applications include pots, pans, vats, kettles, and counters.

3 Month Warranty
Specifications
  • MaterialNylon
  • ColorWhite
  • Handle Length (in)8
  • DescriptionUtility brush with 8" handle, nylon bristles, white
  • Warranty3 month
Reviews
Q&A