Utility Brush, 8.5", Polyester Bristles, Red

Representative image only.

Utility Brush, 8.5", Polyester Bristles, Red

| Item: 8455034

Effective cleaning for a variety of applications

₹2,300.00INR / Each

₹2,300.00INR / Each
Product Description

Key Features

  • Synthetic block is moisture and bacteria proof
  • Hang-up hole provide easy storage
  • Gnarled grip offers safe handling
  • 3 month warranty

More About this Item

The utility brushes come with medium-stif crimped bristles to help carry more water or solution to the cleaning surface. Applications include pots, pans, vats, kettles, and counters.

3 Month Warranty
Specifications
  • MaterialPolyester
  • ColorRed
  • Length (in)8 1/2
  • DescriptionUtility brush, 8.5", polyester bristles, red
  • Warranty3 month
Reviews
Q&A