United Electric E121-2BSB Explosion-Proof Temperature Switch, 30/250F; Remote Mount
Representative image only.
United Electric E121-2BSB Explosion-Proof Temperature Switch, 30/250F; Remote Mount
United Electric | Mfr: E121-2BSB| Item: 3900826
₹290,000.00INR / Each
₹290,000.00INR / Each
Product Description
Key Features
  • 2 year warranty
More About this Item
FEATURES
  • Legendary UE reliability
  • Explosion-proof
  • Class I, Division 1 & 2 (Zone 1); Class II, Div. 1 and 2; Class III
  • Worldwide approvals and certifications
  • Terminal block wiring
  • Single conduit
  • Enclosure design permits safe and easy access for wiring
  • External adjustment via reference dial
  • Die cast aluminum enclosure, epoxy powder coated and gasketed
Two year warranty
Specifications
  • RelaysSPDT
  • Min Temperature (° F)30
  • Max Temperature (° F)250
  • DescriptionExplosion-Proof Temperature Switch, 30/250F; Remote Mount